Assay Details
| Gene Symbol: MPZL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689507.3 | XP_005689564.1 | 2 - 3 | 426 | 70 | |
| GenBank mRNA | KA312204.1 | - | 4 - 5 | 1646 | 70 | |
More Information
Set Membership: |
|

MPZL2
Genomic Map