Assay Details
| Gene Symbol: TMLHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700536.3 | XP_005700593.1 | 7 - 8 | 1262 | 80 | |
| XM_005700537.3 | XP_005700594.1 | 8 - 9 | 1375 | 80 | ||
| XM_013976344.2 | XP_013831798.1 | 6 - 7 | 1025 | 80 | ||
| XM_018045047.1 | XP_017900536.1 | 6 - 7 | 1080 | 80 | ||
More Information
Set Membership: |
|

TMLHE
Genomic Map