Assay Details
| Gene Symbol: LOC102183041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042671.1 | XP_017898160.1 | 12 - 13 | 1349 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102183041| Gene Symbol: LOC102183041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042671.1 | XP_017898160.1 | 12 - 13 | 1349 | 79 | |
Set Membership: |
|