Assay Details
| Gene Symbol: RBAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697901.3 | XP_005697958.1 | 2 - 3 | 1204 | 124 | |
| XM_005697903.3 | XP_005697960.1 | 4 - 5 | 790 | 124 | ||
| GenBank mRNA | JT691567.1 | - | 6 - 7 | 732 | 124 | |
More Information
Set Membership: |
|

RBAK
Genomic Map