Assay Details
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709619.3 | XP_005709676.3 | 8 - 9 | 1049 | 78 | |
| XM_018044429.1 | XP_017899918.1 | 8 - 9 | 1069 | 78 | ||
| XM_018044431.1 | XP_017899920.1 | 8 - 9 | 1069 | 78 | ||
| GenBank mRNA | JO427299.1 | - | 3 - 4 | 1746 | 78 | |
More Information
Set Membership: |
|

RBMX
Genomic Map