Assay Details
| Gene Symbol: RPS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963957.2 | XP_013819411.1 | 3 - 4 | 615 | 120 | |
| GenBank mRNA | JO579102.1 | - | 1 - 2 | 1006 | 120 | |
| KA318754.1 | - | 1 - 2 | 915 | 120 | ||
| Gene Symbol: LOC108638514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066278.1 | XP_017921767.1 | 1 | 342 | 120 | |
More Information
Set Membership: |
|

RPS26
Genomic Map