Assay Details
| Gene Symbol: LOC102184796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964572.2 | XP_013820026.2 | 1 | 713 | 111 | |
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060253.1 | XP_017915742.1 | 2 - 3 | 368 | 111 | |
More Information
Set Membership: |
|

LOC102184796
Genomic Map