Assay Details
| Gene Symbol: LOC102186858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682349.3 | XP_005682406.3 | 2 | 1416 | 73 | |
| XM_018051016.1 | XP_017906505.1 | 13 | 2600 | 73 | ||
More Information
Set Membership: |
|

LOC102186858
Genomic Map