Assay Details
| Gene Symbol: RPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041842.1 | XP_017897331.1 | 4 | 1535 | 108 | |
| XM_018041843.1 | XP_017897332.1 | 4 | 1535 | 108 | ||
| GenBank mRNA | JO581887.1 | - | 4 | 1574 | 108 | |
More Information
Set Membership: |
|

RPL15
Genomic Map