Assay Details
| Gene Symbol: MED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056752.1 | XP_017912241.1 | 7 | 1183 | 115 | |
| GenBank mRNA | JO576416.1 | - | 1 | 212 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056752.1 | XP_017912241.1 | 7 | 1183 | 115 | |
| GenBank mRNA | JO576416.1 | - | 1 | 212 | 115 | |
Set Membership: |
|