Assay Details
| Gene Symbol: LIMS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065840.1 | XP_017921329.1 | 10 | 1726 | 92 | |
| XM_018065841.1 | XP_017921330.1 | 10 | 1742 | 92 | ||
| XM_018065842.1 | XP_017921331.1 | 10 | 1844 | 92 | ||
| XM_018065843.1 | XP_017921332.1 | 10 | 1927 | 92 | ||
| XM_018065844.1 | XP_017921333.1 | 9 | 1596 | 92 | ||
| GenBank mRNA | KA328306.1 | - | 5 | 985 | 92 | |
More Information
Set Membership: |
|

LIMS2
Genomic Map