Assay Details
| Gene Symbol: HAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683211.3 | XP_005683268.2 | 1 | 564 | 96 | |
| GenBank mRNA | JR116213.1 | - | 3 | 813 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683211.3 | XP_005683268.2 | 1 | 564 | 96 | |
| GenBank mRNA | JR116213.1 | - | 3 | 813 | 96 | |
Set Membership: |
|