Assay Details
| Gene Symbol: HOXD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065012.1 | XP_017920501.1 | 1 | 444 | 64 | |
| GenBank mRNA | JT677406.1 | - | 1 | 53 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXD11| Gene Symbol: HOXD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065012.1 | XP_017920501.1 | 1 | 444 | 64 | |
| GenBank mRNA | JT677406.1 | - | 1 | 53 | 64 | |
Set Membership: |
|