Assay Details
| Gene Symbol: JPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709697.3 | XP_005709754.3 | 5 | 1378 | 122 | |
| GenBank mRNA | JO581029.1 | - | 5 | 1417 | 121 | |
More Information
Set Membership: |
|

JPT1
Genomic Map