Assay Details
| Gene Symbol: RBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048809.1 | XP_017904298.1 | 4 | 517 | 106 | |
| XM_018048810.1 | XP_017904299.1 | 5 | 588 | 106 | ||
| GenBank mRNA | JO582117.1 | - | 5 | 619 | 106 | |
More Information
Set Membership: |
|

RBX1
Genomic Map