Assay Details
| Gene Symbol: RRM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983392.2 | XP_008981640.1 | 3 - 4 | 374 | 61 | |
| XM_008983393.2 | XP_008981641.2 | 3 - 4 | 539 | 61 | ||
| XM_017965649.1 | XP_017821138.1 | 4 - 5 | 455 | 61 | ||
More Information
Set Membership: |
|

RRM2B
Genomic Map