Assay Details
| Gene Symbol: ZRANB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756704.4 | XP_002756750.2 | 4 - 5 | 1274 | 76 | |
| XM_008978875.2 | XP_008977123.1 | 3 - 4 | 1166 | 76 | ||
| XM_017963254.1 | XP_017818743.1 | 3 - 4 | 2074 | 76 | ||
More Information
Set Membership: |
|

ZRANB1
Genomic Map