Assay Details
| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744220.3 | XP_002744266.1 | 6 - 7 | 855 | 90 | |
| XM_008985772.2 | XP_008984020.1 | 6 - 7 | 856 | 90 | ||
More Information
Set Membership: |
|

DNAJC18
Genomic Map