Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757284.4 | XP_002757330.1 | 5 - 6 | 3945 | 75 | |
| XM_008979893.2 | XP_008978141.1 | 5 - 6 | 3542 | 75 | ||
| XM_008979896.2 | XP_008978144.1 | 5 - 6 | 3945 | 75 | ||
| XM_008979897.2 | XP_008978145.1 | 5 - 6 | 3945 | 75 | ||

ZNF532
Genomic Map