Assay Details
| Gene Symbol: FAM214B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743058.3 | XP_002743104.1 | 8 - 9 | 2029 | 78 | |
| XM_008998886.1 | XP_008997134.1 | 8 - 9 | 1978 | 78 | ||
| XM_008998893.2 | XP_008997141.1 | 8 - 9 | 1851 | 78 | ||
| XM_008998895.2 | XP_008997143.1 | 8 - 9 | 1780 | 78 | ||
| XM_008998897.1 | XP_008997145.1 | 8 - 9 | 1824 | 78 | ||
| XM_008998903.1 | XP_008997151.1 | 8 - 9 | 1750 | 78 | ||
| XM_008998908.2 | XP_008997156.1 | 8 - 9 | 1723 | 78 | ||
More Information
Set Membership: |
|

FAM214B
Genomic Map