Assay Details
| Gene Symbol: IL5RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257236.1 | NP_001244165.1 | 6 - 7 | 729 | 72 | |
| XM_008982029.2 | XP_008980277.1 | 6 - 7 | 926 | 72 | ||
| XM_017964599.1 | XP_017820088.1 | 6 - 7 | 925 | 72 | ||
| XM_017964600.1 | XP_017820089.1 | 6 - 7 | 925 | 72 | ||
| GenBank mRNA | DQ826672.1 | - | 6 - 7 | 729 | 72 | |
More Information
Set Membership: |
|

IL5RA
Genomic Map