Assay Details
| Gene Symbol: PCYOX1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744353.3 | XP_002744399.1 | 2 - 3 | 364 | 84 | |
| XM_008987650.2 | XP_008985898.1 | 2 - 3 | 334 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008987650.2 transcripts.
Set Membership: |
|

PCYOX1L
Genomic Map