Assay Details
| Gene Symbol: DOLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743369.3 | XP_002743415.1 | 3 - 4 | 374 | 93 | |
| XM_009003779.2 | XP_009002027.1 | 3 - 4 | 386 | 93 | ||
| XM_017976652.1 | XP_017832141.1 | 3 - 4 | 390 | 93 | ||
More Information
Set Membership: |
|

DOLPP1
Genomic Map