Assay Details
| Gene Symbol: GSR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185045.1 | NP_001171974.1 | 1 - 2 | 400 | 88 | |
| GenBank mRNA | EF203010.1 | - | 1 - 2 | 400 | 88 | |
More Information
Set Membership: |
|

GSR
Genomic Map