Assay Details
| Gene Symbol: SESN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746963.3 | XP_002747009.1 | 2 - 3 | 257 | 60 | |
| XM_002746964.4 | XP_002747010.1 | 2 - 3 | 770 | 60 | ||
| XM_017971296.1 | XP_017826785.1 | 2 - 3 | 564 | 60 | ||
More Information
Set Membership: |
|

SESN1
Genomic Map