Assay Details
| Gene Symbol: WASHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759233.3 | XP_002759279.1 | 24 - 25 | 3303 | 63 | |
| XM_017965603.1 | XP_017821092.1 | 23 - 24 | 3153 | 63 | ||
More Information
Set Membership: |
|

WASHC5
Genomic Map