Assay Details
| Gene Symbol: PAFAH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747801.4 | XP_002747847.1 | 6 - 7 | 859 | 67 | |
| XM_008996347.2 | XP_008994595.1 | 7 - 8 | 927 | 67 | ||
More Information
Set Membership: |
|

PAFAH1B1
Genomic Map