Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756202.3 | XP_002756248.2 | 8 - 9 | 1477 | 79 | |
| XM_009009684.2 | XP_009007932.1 | 8 - 9 | 1182 | 79 | ||
| XM_009009685.2 | XP_009007933.1 | 8 - 9 | 1442 | 79 | ||
More Information
Set Membership: |
|

AP3M1
Genomic Map