Assay Details
| Gene Symbol: L3MBTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963881.1 | XP_017819370.1 | 14 - 15 | 1440 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

L3MBTL4| Gene Symbol: L3MBTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963881.1 | XP_017819370.1 | 14 - 15 | 1440 | 106 | |
Set Membership: |
|