Assay Details
| Gene Symbol: RAB21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752762.4 | XP_002752808.1 | 6 - 7 | 782 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB21| Gene Symbol: RAB21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752762.4 | XP_002752808.1 | 6 - 7 | 782 | 116 | |
Set Membership: |
|