Assay Details
| Gene Symbol: RELCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757255.3 | XP_002757301.1 | 12 - 13 | 2044 | 74 | |
| XM_008979915.2 | XP_008978163.1 | 13 - 14 | 2110 | 74 | ||
| XM_008979916.2 | XP_008978164.1 | 13 - 14 | 2110 | 74 | ||
| XM_008979917.2 | XP_008978165.1 | 12 - 13 | 1829 | 74 | ||
| XM_008979918.2 | XP_008978166.1 | 12 - 13 | 2038 | 74 | ||
| XM_008979919.2 | XP_008978167.1 | 11 - 12 | 1757 | 74 | ||
| XM_017963524.1 | XP_017819013.1 | 13 - 14 | 2110 | 74 | ||
More Information
Set Membership: |
|

RELCH
Genomic Map