Assay Details
Gene Symbol: RBM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747709.3 | XP_002747755.1 | 1 - 2 | 382 | 79 | |
XM_008996161.1 | XP_008994409.1 | 1 - 2 | 382 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
