Assay Details
| Gene Symbol: DEPTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759264.4 | XP_002759310.1 | 5 - 6 | 973 | 67 | |
| XM_017965621.1 | XP_017821110.1 | 5 - 6 | 973 | 67 | ||
More Information
Set Membership: |
|

DEPTOR
Genomic Map