Assay Details
| Gene Symbol: TGM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995639.1 | XP_008993887.1 | 7 - 8 | 1043 | 60 | |
| XM_008995640.1 | XP_008993888.1 | 7 - 8 | 1043 | 60 | ||
More Information
Set Membership: |
|

TGM6
Genomic Map