Assay Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750859.4 | XP_002750905.2 | 1 - 2 | 397 | 64 | |
| XM_009001287.2 | XP_008999535.1 | 1 - 2 | 434 | 64 | ||
More Information
Set Membership: |
|

TMEM59
Genomic Map