Assay Details
| Gene Symbol: VPS52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994235.2 | XP_008992483.1 | 13 - 14 | 1713 | 74 | |
| XM_008994236.1 | XP_008992484.1 | 12 - 13 | 1557 | 74 | ||
| XM_017971010.1 | XP_017826499.1 | 11 - 12 | 1521 | 74 | ||
More Information
Set Membership: |
|

VPS52
Genomic Map