Assay Details
| Gene Symbol: MTSS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986187.2 | XP_008984435.2 | 11 - 12 | 1716 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTSS1L| Gene Symbol: MTSS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986187.2 | XP_008984435.2 | 11 - 12 | 1716 | 63 | |
Set Membership: |
|