Assay Details
Gene Symbol: MATR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205240.1 | NP_001192169.1 | 8 - 9 | 1772 | 102 | |
XM_008984987.1 | XP_008983235.1 | 8 - 9 | 1763 | 102 | ||
XM_008984991.1 | XP_008983239.1 | 11 - 12 | 1898 | 102 | ||
XM_008984997.1 | XP_008983245.1 | 8 - 9 | 1855 | 102 | ||
XM_008985004.1 | XP_008983252.1 | 8 - 9 | 1701 | 102 | ||
XM_008985011.2 | XP_008983259.1 | 8 - 9 | 1642 | 102 | ||
XM_008985017.1 | XP_008983265.1 | 7 - 8 | 794 | 102 | ||
XM_008985022.2 | XP_008983270.1 | 7 - 8 | 799 | 102 | ||
XM_017964672.1 | XP_017820161.1 | 12 - 13 | 2052 | 102 | ||
XM_017964682.1 | XP_017820171.1 | 10 - 11 | 882 | 102 | ||
XM_017964684.1 | XP_017820173.1 | 9 - 10 | 851 | 102 | ||
XM_017964686.1 | XP_017820175.1 | 6 - 7 | 1588 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
