Assay Details
| Gene Symbol: PKNOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986751.2 | XP_008984999.1 | 8 - 9 | 1054 | 86 | |
| XM_017967199.1 | XP_017822688.1 | 8 - 9 | 2334 | 86 | ||
| XM_017967200.1 | XP_017822689.1 | 8 - 9 | 1051 | 86 | ||
| GenBank mRNA | EF215161.1 | - | 2 - 3 | 172 | 86 | |
More Information
Set Membership: |
|

PKNOX1
Genomic Map