Assay Details
| Gene Symbol: EFEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757741.2 | XP_002757787.2 | 7 - 8 | 1302 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFEMP1| Gene Symbol: EFEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757741.2 | XP_002757787.2 | 7 - 8 | 1302 | 70 | |
Set Membership: |
|