Assay Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763145.3 | XP_002763191.1 | 2 - 3 | 998 | 81 | |
| XM_008989720.2 | XP_008987968.1 | 2 - 3 | 821 | 81 | ||
More Information
Set Membership: |
|

RNF128
Genomic Map