Assay Details
| Gene Symbol: ETV6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752140.3 | XP_002752186.1 | 5 - 6 | 1262 | 64 | |
| XM_009003317.2 | XP_009001565.1 | 5 - 6 | 1207 | 64 | ||
| XM_017975822.1 | XP_017831311.1 | 5 - 6 | 1620 | 64 | ||
| XM_017975823.1 | XP_017831312.1 | 6 - 7 | 1428 | 64 | ||
| XM_017975824.1 | XP_017831313.1 | 6 - 7 | 1395 | 64 | ||
| XM_017975825.1 | XP_017831314.1 | 4 - 5 | 862 | 64 | ||
| XM_017975826.1 | XP_017831315.1 | 4 - 5 | 2070 | 64 | ||
More Information
Set Membership: |
|

ETV6
Genomic Map