Assay Details
| Gene Symbol: RUNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994524.1 | XP_008992772.1 | 2 - 3 | 608 | 86 | |
| XM_017971129.1 | XP_017826618.1 | 2 - 3 | 608 | 86 | ||
| XM_017971130.1 | XP_017826619.1 | 608 | 86 | |||
More Information
Set Membership: |
|

RUNX2
Genomic Map