Assay Details
| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757550.3 | XP_002757596.1 | 3 - 4 | 320 | 103 | |
| XM_008980404.1 | XP_008978652.1 | 3 - 4 | 315 | 103 | ||
| XM_017964194.1 | XP_017819683.1 | 3 - 4 | 421 | 103 | ||
| XM_017964195.1 | XP_017819684.1 | 2 - 3 | 554 | 103 | ||
| XM_017964196.1 | XP_017819685.1 | 3 - 4 | 421 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008980404.1, XM_017964195.1, XM_017964196.1 transcripts.
Set Membership: |
|

TMEM150A
Genomic Map