Assay Details
| Gene Symbol: OAZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289928.1 | NP_001276857.1 | 4 - 5 | 648 | 61 | |
| NM_001301337.1 | NP_001288266.1 | 4 - 5 | 645 | 61 | ||
| NR_125741.1 | 6 - 7 | 852 | 61 | |||
| NR_125742.1 | 6 - 7 | 806 | 61 | |||
| NR_125743.1 | 5 - 6 | 758 | 61 | |||
| NR_125744.1 | 3 - 4 | 490 | 61 | |||
| GenBank mRNA | HX600531.1 | - | 6 - 7 | 650 | 61 | |
| HX601182.1 | - | 5 - 6 | 722 | 61 | ||
| HX602668.1 | - | 3 - 4 | 454 | 61 | ||
| HX662041.1 | - | 4 - 5 | 649 | 61 | ||
More Information
Set Membership: |
|

OAZ2
Genomic Map