Assay Details
| Gene Symbol: VPS33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753118.3 | XP_002753164.1 | 7 - 8 | 1082 | 91 | |
| XM_009004823.2 | XP_009003071.1 | 7 - 8 | 1082 | 91 | ||
More Information
Set Membership: |
|

VPS33A
Genomic Map