Assay Details
| Gene Symbol: HPSE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756506.2 | XP_002756552.1 | 11 - 12 | 1684 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPSE2| Gene Symbol: HPSE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756506.2 | XP_002756552.1 | 11 - 12 | 1684 | 69 | |
Set Membership: |
|