Assay Details
| Gene Symbol: IMPACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979626.2 | XP_008977874.1 | 8 - 9 | 803 | 89 | |
| XM_008979627.2 | XP_008977875.1 | 8 - 9 | 803 | 89 | ||
| XM_008979628.2 | XP_008977876.1 | 8 - 9 | 803 | 89 | ||
| XM_008979630.2 | XP_008977878.1 | 8 - 9 | 803 | 89 | ||
| XM_017963927.1 | XP_017819416.1 | 8 - 9 | 803 | 89 | ||
| XM_017963929.1 | XP_017819418.1 | 8 - 9 | 1554 | 89 | ||
| XM_017963930.1 | XP_017819419.1 | 6 - 7 | 556 | 89 | ||
More Information
Set Membership: |
|

IMPACT
Genomic Map