Assay Details
| Gene Symbol: CDK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744895.4 | XP_002744941.1 | 8 - 9 | 766 | 68 | |
| XM_017969885.1 | XP_017825374.1 | 9 - 10 | 881 | 68 | ||
| XM_017969886.1 | XP_017825375.1 | 8 - 9 | 621 | 68 | ||
| XM_017969887.1 | XP_017825376.1 | 9 - 10 | 778 | 68 | ||
More Information
Set Membership: |
|

CDK7
Genomic Map