Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760305.2 | XP_002760351.1 | 2 - 3 | 689 | 62 | |
| XM_008984913.1 | XP_008983161.1 | 2 - 3 | 680 | 62 | ||
More Information
Set Membership: |
|

RGS4
Genomic Map